Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MMP26 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580820
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human MMP-26/Endometase.
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. The encoded protein degrades type IV collagen, fibronectin, fibrinogen, casein, vitronectin, alpha 1-antitrypsin, alpha 2-macroglobulin, and insulin-like growth factor-binding protein 1, and activates MMP9 by cleavage. The protein differs from most MMP family members in that it lacks a conserved C-terminal protein domain.
Specifications
MMP26 | |
Polyclonal | |
Unconjugated | |
MMP26 | |
Endometase; matrilysin 2; matrilysin-2; matrix metallo protease; matrix metallopeptidase 26; matrix metalloproteinase 26; matrix metalloproteinase-26; MGC126590; MGC126592; MMP; MMP26; MMP-26; MMPs | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
56547 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
1 mg/mL | |
PBS with no preservative | |
Q9NRE1 | |
MMP26 | |
Recombinant Human MMP-26/Endometase Protein (Thr90-Pro261). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction