Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP26 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 180871AP150UL
Description
This antibody is specific to MMP26.
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP′s are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. The encoded protein degrades type IV collagen, fibronectin, fibrinogen, casein, vitronectin, alpha 1-antitrypsin, alpha 2-macroglobulin, and insulin-like growth factor-binding protein 1, and activates MMP9 by cleavage. The protein differs from most MMP family members in that it lacks a conserved C-terminal protein domain.Specifications
MMP26 | |
Polyclonal | |
Unconjugated | |
MMP26 | |
Endometase, Matrilysin 2, matrix metallopeptidase 26, Matrix metalloproteinase 26, MMP 26, MMP26 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56547 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NRE1 | |
MMP26 | |
MMP26 Fusion Protein Ag12806 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction