Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP28 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309440100UL
Description
MMP28 Polyclonal specifically detects MMP28 in Human samples. It is validated for Western Blot.Specifications
MMP28 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
EC 3.4.24.-, EPILYSIN, matrix metallopeptidase 28, matrix metalloproteinase 28, MM28, MMP25, MMP-28MMP-25, putative MMP28 gene product | |
The immunogen is a synthetic peptide directed towards the N-terminal region of human MMP28 (NP_116568.1). Peptide sequence LDAQPAERGGQELRKEAEAFLEKYGYLNEQVPKAPTSTRFSDAIRAFQWV | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
79148 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction