Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551003

Catalog No. PA551003


Add to cart

Description

Description

MMP8 Polyclonal Antibody detects endogenous levels of MMP8 protein.

Can degrade fibrillar type I, II, and III collagens.
Specifications

Specifications

MMP8
Polyclonal
Unconjugated
MMP8
Liquid
MMP8
Synthesized peptide derived from the C-terminal region of human MMP-8.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
ELISA, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
P22894, O70138, O88766
CLG1, HNC, PMNL-CL
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
17394, 4317, 63849
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit