Learn More
MMPIP hydrochloride, Tocris
Potent, allosteric mGlu7-selective antagonist
Manufacturer:  R&D Systems 2963/50
Quantity 50mg
 View more versions of this product
Catalog No. 29-635-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>98%
369.8
MMPIP hydrochloride
C19H15N3O3·HCl
6-(4-Methoxyphenyl)-5-methyl-3-(4-pyridinyl)-isoxazolo[4, 5-c]pyridin-4(5H)-one hydrochloride
Specifications
479077-02-6
C19H15N3O3·HCl
50mg
MMPIP hydrochloride
>98%
369.8
6-(4-Methoxyphenyl)-5-methyl-3-(4-pyridinyl)-isoxazolo[4, 5-c]pyridin-4(5H)-one hydrochloride
Safety and Handling
Recommended Storage : Desiccate at Room Temperature