Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMPIP hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Description
Potent allosteric mGlu7-selective receptor antagonist. Inhibits agonist-induced intracellular calcium mobilization and cAMP accumulation (IC50 values are 26 and 610 nM). Displays intrinsic activity; increases forskolin-induced cAMP accumulation in the absence of an agonist (IC50 = 15 nM), indicating inverse agonist activity.
Specifications
Quantity | 10 mg |
Formula Weight | 369.8 |
Percent Purity | >98% |
Chemical Name or Material | MMPIP hydrochloride |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction