Learn More
Tocris Bioscience™ MMPX Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Tocris Bioscience™ 0552/50
 View more versions of this product
Catalog No. 05-525-0
Add to cart
This item is not returnable. View return policy
Description
Description
Specific inhibitor of calmodulin-sensitive cyclic GMP phosphodiesterase (IC50 = 5.2 µM).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
8-Methoxymethyl-3-isobutyl-1-methylxanthine
C12H18N4O3
50mg
78033-08-6
Observed MW: 266.3
Soluble to 100 mM in ethanol
Safety and Handling
Safety and Handling
Recommended Storage : Store at RT