Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MMPX Available on GSA/VA Contract for Federal Government customers only.

$163.00 - $683.00

Specifications

Formula Weight Observed MW: 266.3
Chemical Name or Material 8-Methoxymethyl-3-isobutyl-1-methylxanthine
Molecular Formula C12H18N4O3
CAS 78033-08-6
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
05-525-0 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Tocris Bioscience™
0552/50
50mg Each for $683.00
Add to cart
 
05-521-0 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Tocris Bioscience™
0552/10
10mg Each for $163.00
Add to cart
 
Description

Description

Specific inhibitor of calmodulin-sensitive cyclic GMP phosphodiesterase (IC50 = 5.2 µM).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Observed MW: 266.3
8-Methoxymethyl-3-isobutyl-1-methylxanthine
C12H18N4O3
78033-08-6
Safety and Handling

Safety and Handling

Recommended Storage : Store at RT

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit