Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMRN2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548501

Catalog No. PA548501

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Inhibits endothelial cells motility and acts as a negative regulator of angiogenesis; it downregulates KDR activation by binding VEGFA.
Specifications

Specifications

MMRN2
Polyclonal
PBS with 0.09% sodium azide
Q9H8L6
Multimerin-2, EMILIN-3, Elastin microfibril interface located protein 3, Elastin microfibril interfacer 3, EndoGlyx-1 p125/p140 subunit, MMRN2, EMILIN3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79812
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MMRN2
Liquid
MMRN2
KLH conjugated synthetic peptide between 768-797 amino acids from the C-terminal region of human MMRN2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit