Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMS19 like protein Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $487.50

Specifications

Antigen MMS19 like protein
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095639
SDP
View Documents
Novus Biologicals
NBP30379920UL
20 μL
Each for $181.90
Only null left
Add to Cart
 
NB095638
SDP
View Documents
Novus Biologicals
NBP303799100UL
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

MMS19 like protein Polyclonal specifically detects MMS19 like protein in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

MMS19 like protein
Polyclonal
Purified
RUO
PBS with 50% glycerol, pH7.3.
64210
IgG
Affinity purified
Western Blot
Unconjugated
Rabbit
Mouse
FLJ34167, MET18, MET18 homolog, MMS19 nucleotide excision repair homolog (S. cerevisiae), S. cerevisiae)
Recombinant fusion protein containing a sequence corresponding to amino acids 761-1030 of human MMS19 like protein (NP_071757.4).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.