Learn More
Invitrogen™ MMS19 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596306
Description
Immunogen sequence: FAGLLNKHPA GQQLDEFLQL AVDKVEAGLG SGPCRSQAFT LLLWVTKALV LRYHPLSSCL TARLMGLLSD PELGPAAADG FSLLMSDCTD VLTRAGHAEV RIMFRQRFFT DNVPALVQGF HAAPQDVKPN YLKGLSHVLN RLPKPVLLPE LPTLLSLLLE ALSCPDCVVQ LSTLSCLQPL LLEAPQVMSL HVDTLVTKFL NLSSSPSMAV RIAALQCMHA LTRLPTPVLL PYKPQVIRAL AKPLDDKKRL VRKEAVSARG EWFLLGSPGS; Positive Samples: Mouse thymus; Cellular Location: Cytoplasm, Nucleus, cytoskeleton, spindle.
Key component of the cytosolic iron-sulfur protein assembly (CIA) complex, a multiprotein complex that mediates the incorporation of iron-sulfur cluster into apoproteins specifically involved in DNA metabolism and genomic integrity. In the CIA complex, MMS19 acts as an adapter between early-acting CIA components and a subset of cellular target iron-sulfur proteins such as ERCC2/XPD, FANCJ and RTEL1, thereby playing a key role in nucleotide excision repair (NER) and RNA polymerase II (POL II) transcription. As part of the mitotic spindle-associated MMXD complex, plays a role in chromosome segregation, probably by facilitating iron-sulfur cluster assembly into ERCC2/XPD. Indirectly acts as a transcriptional coactivator of estrogen receptor (ER), via its role in iron-sulfur insertion into some component of the TFIIH-machinery.
Specifications
MMS19 | |
Polyclonal | |
Unconjugated | |
MMS19 | |
2410001K24Rik; 2610042O15Rik; AI316855; C79368; C86341; hMMS19; homolog of yeast MMS19; incisor protein; MET18; MET18 homolog; Mms19; MMS19 (MET18 S. cerevisiae); MMS19 cytosolic iron-sulfur assembly component; MMS19 homolog, cytosolic iron-sulfur assembly component; MMS19 nucleotide excision repair homolog; MMS19 nucleotide excision repair homolog (S. cerevisiae); MMS19 nucleotide excision repair protein homolog; MMS19 nucleotide excision repair-like; Mms19l; MMS19-like (MET18 homolog, S. cerevisiae); MMS19-like protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
72199 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.03 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9D071 | |
MMS19 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 761-1030 of human MMS19 (NP_0717574). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.