Tocris™ MN 64

Potent and selective tankyrase inhibitor

Manufacturer: Tocris™ 5084/50

 View more versions of this product

Catalog No. 50-845-0R

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors MN 64
    Purity >98
    Molecular Weight 264.32
    Quantity 50mg
    Product Type MN 64

    Potent and selective tankyrase (TNKS) inhibitor (IC50 values are 6 and 72nM for TNKS1 and TNKS2 respectively). Has no significant effect on PARP-1 or PARP-2 (IC50 >19µM). Inhibits WNT/β-catenin signaling-dependent Super-TopFlash (STF) luciferase activity.

    • Chemical Name: 2-[4-(1-Methylethyl)phenyl]-4H-1-benzopyran-4-one
    • Formula: C18H16O2
    • CAS Number: 92831-11-3
    • Solubility: Soluble to 100mM in DMSO and to 100mM in ethanol
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit