Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599238
Description
Antibody detects endogenous levels of total MN1.
Meningioma 1 (MN1) contains two sets of CAG repeats. It is disrupted by a balanced translocation (4; 22) in a meningioma, and its inactivation may contribute to meningioma 32 pathogenesis.
Specifications
MN1 | |
Polyclonal | |
Unconjugated | |
MN1 | |
AA003644; AA009236; dJ353E16.2; meningioma (disrupted in balanced translocation) 1; meningioma (translocation balanced); meningioma 1; meningioma chromosome region 1; MGCR; MGCR1; MGCR1-PEN; MN1; MN1 proto-oncogene, transcriptional regulator; probable tumor suppressor protein MN1; RGD1565571; Transcriptional activator MN1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
4330, 433938 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
D3YWE6, Q10571 | |
MN1 | |
A synthesized peptide derived from human MN1(Accession Q10571), corresponding to amino acid residues A831-G881. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction