Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MNI caged kainic acid, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
This item is not returnable. View return policy
1315378-75-6
C19H23N3O6
389.408
IQGCUMUIYXHZOT-AHIWAGSCSA-N
mni caged kainic acid,2s,3s,4s-carboxy-4-1-methylethenyl-3-pyrrolidineacetic acid 4-methoxy-7-nitro-1h-indolinyl amide
57414719
(2S,3S,4S)-3-[2-(4-methoxy-7-nitro-2,3-dihydroindol-1-yl)-2-oxoethyl]-4-prop-1-en-2-ylpyrrolidine-2-carboxylic acid
CC(=C)C1CNC(C1CC(=O)N2CCC3=C(C=CC(=C32)[N+](=O)[O-])OC)C(=O)O
This item is not returnable. View return policy
1315378-75-6
C19H23N3O6
389.408
IQGCUMUIYXHZOT-AHIWAGSCSA-N
mni caged kainic acid,2s,3s,4s-carboxy-4-1-methylethenyl-3-pyrrolidineacetic acid 4-methoxy-7-nitro-1h-indolinyl amide
57414719
(2S,3S,4S)-3-[2-(4-methoxy-7-nitro-2,3-dihydroindol-1-yl)-2-oxoethyl]-4-prop-1-en-2-ylpyrrolidine-2-carboxylic acid
CC(=C)C1CNC(C1CC(=O)N2CCC3=C(C=CC(=C32)[N+](=O)[O-])OC)C(=O)O

Caged kainic acid

Kainic acid (Cat.No. 0222) caged with the photosensitive 4-methoxy-7-nitroindolinyl group. Generates large inward currents at resting membrane potential upon wide field photolysis in Purkinje neurons. Selectively photoactivates kainate receptors when used in conjunction with an AMPA antagonist, such as GYKI 53655 (Cat.No. 2555). Suitable for uncaging with 300-380 nm excitation. Exhibits rapid uncaging rate relative to ionotropic glutamate receptor activation. Also suitable for two-photon uncaging experiments at 720 nm.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 389.4
Percent Purity >99%
Chemical Name or Material MNI caged kainic acid

Recommended Storage : Store at -20°C