Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MNI-caged-NMDA, Tocris Bioscience™

Catalog Number 222410
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
1227675-52-6
C14H17N3O6
323.305
MUCRWOILMOHLGD-SECBINFHSA-N
mni-caged-nmda, 4-methoxy-7-nitroindolinyl-caged n-methyl-d-aspartic acid, r-?-methylamino-2,3-dihydro-4-methoxy-7-nitro-?-oxo-1h-indole-1-butanoic acid, alphar-2,3-dihydro-4-methoxy-alpha-methylamino-7-nitro-gamma-oxo-1h-indole-1-butanoic acid
57369294
(2R)-4-(4-methoxy-7-nitro-2,3-dihydroindol-1-yl)-2-(methylamino)-4-oxobutanoic acid
CNC(CC(=O)N1CCC2=C(C=CC(=C21)[N+](=O)[O-])OC)C(=O)O
This item is not returnable. View return policy
1227675-52-6
C14H17N3O6
323.305
MUCRWOILMOHLGD-SECBINFHSA-N
mni-caged-nmda, 4-methoxy-7-nitroindolinyl-caged n-methyl-d-aspartic acid, r-?-methylamino-2,3-dihydro-4-methoxy-7-nitro-?-oxo-1h-indole-1-butanoic acid, alphar-2,3-dihydro-4-methoxy-alpha-methylamino-7-nitro-gamma-oxo-1h-indole-1-butanoic acid
57369294
(2R)-4-(4-methoxy-7-nitro-2,3-dihydroindol-1-yl)-2-(methylamino)-4-oxobutanoic acid
CNC(CC(=O)N1CCC2=C(C=CC(=C21)[N+](=O)[O-])OC)C(=O)O

Caged NMDA

NMDA (Cat.No. 0114) caged with the photosensitive 4-methoxy-7-nitroindolinyl group. Stoichiometrically releases chirally pure NMDA. Suitable for uncaging with 300-380 nm excitation. Improved localization can be achieved by uncaging at 405 nm (with mM concentration of caged compound in the bath solution). Exhibits rapid uncaging rate relative to ionotropic glutamate receptor activation.
Quantity 10 mg
Formula Weight 323.3
Percent Purity >99%
Chemical Name or Material MNI-caged-NMDA
Recommended Storage Store at -20°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit