Learn More
Tocris Bioscience™ MNITMT
Manufacturer:  Tocris Bioscience™ 2492/50
 View more versions of this product
Catalog No. 24-925-0
Add to cart
This item is not returnable. View return policy
Description
Description
Novel, non-toxic immunosuppressive agent. Inhibits human mixed lymphocyte reaction in vitro (ED 50 = 2.7 µM) and prolongs skin graft survival in mice. Produces a 98.1% reduction in antibody response following oral administration in rabbits. Previously sold as Cat. No. 0564.
Specifications
Specifications
4-Methyl-3-[(1-methyl-4-nitro-1H-imidazol-5-yl)thio]-4H-1,2,4-triazole
>99%
Observed MW: 240.24
Soluble to 20mM in DMSO
177653-76-8
C7H8N6O2S
50mg
Safety and Handling
Safety and Handling
Recommended Storage : Store at +4°C