Learn More
Tocris Bioscience™ MNITMT
$149.00 - $609.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
24-925-0 View Documents Tocris Bioscience™
2492/50
Each for $609.00
Add to cart
24-921-0 View Documents Tocris Bioscience™
2492/10
10mg Each for $149.00
Add to cart
Description
Description
Novel, non-toxic immunosuppressive agent. Inhibits human mixed lymphocyte reaction in vitro (ED50 = 2.7 µM) and prolongs skin graft survival in mice. Produces a 98.1% reduction in antibody response following oral administration in rabbits.
Previously sold as Cat. No. 0564.
Safety and Handling
Safety and Handling
Recommended Storage : Store at +4°C