Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MNK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

$395.30

Specifications

Antigen MNK1
Concentration 0.15 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Conjugate Unconjugated
Format Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-172-6051
Encompass_Preferred
View Documents
Proteintech Group IncSupplier Diversity Partner
101361AP150UL
150 μL
Each for $395.30
Add to cart
 
Description

Description

MKNK1 encodes a predicted 424-amino acid protein containing a sequence typical of the catalytic domain of serine/threonine kinases and a putative N-terminal nuclear localization signal sequence. C-terminal region of MKNK1 was phosphorylated and activated in vivo and in vitro by ERK1 and p38 MAP kinases, but not by JNK/SAPK. The activated ERK is essential for the activation of MKNK1 in insect cells. MKNK1 was activated following stimulation of HeLa cells with a variety of extracellular stimuli. MNK1 bind tightly to the growth factor-regulated MAP kinases, Erk1 and Erk2, and also binds strongly to the stress-activated kinase, p38.
Specifications

Specifications

MNK1
Immunocytochemistry, Immunofluorescence, Western Blot
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Polyclonal
-20°C
0.15 mg/mL
Unconjugated
Rabbit
Human, Mouse, Rat
IgG
Antigen Affinity Chromatography
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit