Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MOB4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571430
Description
MOB4 Polyclonal Antibody for Western Blot
This gene was identified based on its similarity with the mouse counterpart. Studies of the mouse counterpart suggest that the expression of this gene may be regulated during oocyte maturation and preimplantation following zygotic gene activation. Alternatively spliced transcript variants encoding distinct isoforms have been observed. Naturally occurring read-through transcription occurs between this locus and the neighboring locus HSPE1.
Specifications
MOB4 | |
Polyclonal | |
Unconjugated | |
MOB4 | |
2610109B12Rik; 2C4D; CGI-95; Class II mMOB1; MOB family member 4, phocein; MOB1; Mob1 homolog 3; MOB1, Mps One Binder kinase activator-like 3; mob3; MOB4; Mobkl1; Mobkl3; MOB-like protein phocein; mps one binder kinase activator-like 3; Phocein; phocein, Mob-like protein; PHOCN; PREI3; Preimplantation protein 3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
25843 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y3A3 | |
MOB4 | |
synthetic peptide directed towards the N terminal region of human MOB4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction