Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MOB4A Mouse anti-Human, Mouse, Rat, DyLight 405, Clone: OTI3G9, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272744V
Description
MOB4A Monoclonal specifically detects MOB4A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MOB4A | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MOB1B | |
Full length recombinant protein of human MOB4A (NP_775739) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI3G9 | |
Western Blot, Immunohistochemistry | |
MATS2, MGC33910, Mob1 homolog 1A, MOB1, Mps One Binder kinase activator-like 1A (yeast), Mob1A, Mob1Bmps one binder kinase activator-like 1A, MOB4Amob1A, Protein Mob4A | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
92597.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title