Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOBKL1B Mouse anti-Human, Clone: 3E7B2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP261874

Catalog No. NBP2-61874


Add to cart

Description

Description

MOBKL1B Monoclonal antibody specifically detects MOBKL1B in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry.
Specifications

Specifications

MOBKL1B
3E7B2
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000
Purified
MOB1A
Purified recombinant fragment of human MOB1A (AA:1-216) expressed in E. Coli.
25 kDa
0.1 ml
Primary
55233
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
PBS with 0.05% Sodium Azide
C2orf6, chromosome 2 open reading frame 6, FLJ10788, FLJ11595, MATS1, MOB1, Mob1 alpha, Mob1 homolog 1B, MOB1, Mps One Binder kinase activator-like 1B (yeast), mob1A, Mob4B, MOBK1B, mps one binder kinase activator-like 1B, Protein Mob4B
Mouse
IgG1
Protein G purified
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit