Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOBKL2B Mouse anti-Human, Clone: OTI1F9, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA501557

Catalog No. 50-167-2693


Add to cart

Description

Description

The protein encoded by this gene shares similarity with the yeast Mob1 protein. Yeast Mob1 binds Mps1p, a protein kinase essential for spindle pole body duplication and mitotic checkpoint regulation. This gene is located on the opposite strand as the interferon kappa precursor (IFNK) gene.
Specifications

Specifications

MOBKL2B
Monoclonal
0.49 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q86TA1
C9orf35, MOB1D, MOBKL2B
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79817
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI1F9
Unconjugated
MOB3B
Liquid
MOB3B
Full length human recombit protein of human MOBKL2B produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit