Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MOBKL2B Mouse anti-Human, Mouse, Rat, HRP, Clone: OTI6C6, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272748H
Description
MOBKL2B Monoclonal specifically detects MOBKL2B in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
MOBKL2B | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
MOB3B | |
Full length recombinant protein of human MOBKL2B (NP_079037) produced in HEK293T cells. | |
0.1 mL | |
79817.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI6C6 | |
Western Blot | |
Em:AL163192.1, FLJ13204, FLJ23916, Mob1 homolog 2b, MOB1, Mps One Binder kinase activator-like 2B (yeast), MOB3BMGC32960, monopolar spindle 1 binding, MOB1, domain containing, mps one binder kinase activator-like 2B, Protein Mob3B | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title