Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOCS2 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen MOCS2
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095963
View Documents
Novus Biologicals
NBP304485100UL
100 μg
Each for $320.71
Add to cart
 
NB095964
View Documents
Novus Biologicals
NBP30448520UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

MOCS2 Polyclonal antibody specifically detects MOCS2 in Mouse samples. It is validated for Western Blot
Specifications

Specifications

MOCS2
Unconjugated
Rabbit
Mouse
EC 2.-, MCBPE, MOCO1-A, MOCO1-B, MOCO1MOCS2B, MOCS2A, molybdenum cofactor biosynthesis protein E, molybdenum cofactor synthesis 2, Molybdenum cofactor synthesis protein 2 large subunit, Molybdenum cofactor synthesis protein 2 small subunit, Molybdenum cofactor synthesis protein 2A, Molybdenum cofactor synthesis protein 2B, molybdopterin synthase catalytic subunit, molybdopterin synthase sulfur carrier subunit, Molybdopterin-synthase large subunit, Molybdopterin-synthase small subunit, MPT synthase large subunit, MPTS, Sulfur carrier protein MOCS2A
Recombinant fusion protein containing a sequence corresponding to amino acids 1-188 of human MOCS2 (NP_004522.1).
Polyclonal
Affinity purified
Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
4338
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit