Learn More
Sentry Air Systems Model 300 Pre-Filter Pads – 12 Pack
SDP
Supplier:  Sentry Air Systems SS300PFP MODEL 300
Encompass
Pack of 12 Pre-Filter Pads for Model 300 units with MERV 8 Efficiency Rating. These pre-filter pad replacements help the filtration system capture and remove harmful particulates emitted from different processes. This pack fits all Model 300 units including ductless fume hoods, portable clean rooms, and fume extractors. The pre-filter is used in conjunction with a Model 300 HEPA, ASHRAE, ULPA, Activated Carbon, or specialty-blended filter.
Catalog No. NC1716441
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.