Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Modified Methyl Orange Indicator, 0.01% Methyl Orange-Xylene Cyanole, Reagents
Click to view available options
Quantity:
120 mL
1 L
4 L
Specifications
Specifications
Chemical Name or Material | Modified Methyl Orange Indicator, 0.01% Methyl Orange-Xylene Cyanole |
CAS | 7732-18-5, 2650-17-1 |
Boiling Point | 100°C |
Synonym | Methyl Orange-Xylene Cyanole |
Percent Purity | 0.01% |
Grade | Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1 |
Color | Blue to Green |
Quantity | 4 L |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction