Learn More
Modified Methyl Orange Indicator, 0.1% Methyl Orange-Xylene Cyanole, Reagents
Specifications
Specifications
| Chemical Name or Material | Modified Methyl Orange Indicator, 0.1% Methyl Orange-Xylene Cyanole |
| Boiling Point | 100°C |
| CAS | 7732-18-5, 2650-17-1 |
| Synonym | Methyl Orange-Xylene Cyanole |
| Percent Purity | 0.1% |
| Grade | Reagent |
| Packaging | Poly Bottle |
| Specific Gravity | 1.001 |
| Color | Blue to Green |
| Quantity | 1 L |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.