Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Modular Blocks and Accessories: PROMO Products for Science Education

Constructed from solid anodized aluminum block.

Specifications

For Use With (Equipment) Dry block heater
Height (Metric) 5.1 cm
Length (Metric) 9.5 cm
Material Aluminum
View More Specs

Alert:

To avoid possible electrical hazard, DO NOT fill well or block with water or other liquids. Units are designed as a dry bath/incubator.


Products 8
Catalog Number Mfr. No. Depth (Metric) Diameter (Metric) Well Type Sample Type Price Quantity & Availability  
Catalog Number Mfr. No. Depth (Metric) Diameter (Metric) Well Type Sample Type Price Quantity & Availability  
S38776PMO Products for Science Education View Documents OHAUS™
30400168PMO
2.9 cm Centrifuge Tube Block 50mL Tube This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
S38777PMO Products for Science Education View Documents OHAUS™
30400172PMO
1.71 cm Centrifuge Tube Block 15mL Tube This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
S38778PMO Products for Science Education View Documents OHAUS™
30400164PMO
Titer Plate Block Titer plate This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
S38779PMO Products for Science Education View Documents OHAUS™
30400159PMO
1.1 cm Microcentrifuge Tube Block 1.5mL Tube This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
S38772PMO Products for Science Education View Documents OHAUS™
30400246PMO
Temperature Probe Kit This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
S38773PMO Products for Science Education View Documents OHAUS™
30400185PMO
1.97 cm Vial Block 19mm Vial This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
S38774PMO Products for Science Education View Documents OHAUS™
30400153PMO
1.75 cm Test Tube Block 15/16mm Tube This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
S38775PMO Products for Science Education View Documents OHAUS™
30400151PMO
1.07 cm Test Tube Block 10mm Tube This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Modular blocks are sold separately from heaters.

  • Close contact of tubes-to-block-walls allow for maximum heat retention
  • Each block has a thermometer well for measuring block temperature

Specifications

Specifications

Dry block heater
5.1 cm
9.5 cm
Aluminum
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit