Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Moesin Ab-1, Mouse Monoclonal Antibody

Deliver accurate, reproducible results with the Epredia™ Moesin Ab-1, Mouse Monoclonal Antibody.

Supplier:  Epredia™ MS727R1

Catalog No. MS727R1

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

Moesin, a member of the talin-4.1 superfamily, is a linking protein of the submembraneous actin cytoskeleton. It is expressed in macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. It is involved in cell adhesion, migration, and organization of cell surface structures.

Host Species: Mouse

Clone: 38/87

Isotype: IgG1/κ

Species Reactivity: Human, Cow and Rat. Others not known.

Epitope: Not determined

Immunogen: Moesin purified from cow uterus

Molecular Weight: 78kDa moesin and 80kDa radixin

Positive Control: Raji cells; Placenta

Cellular Localization: Membrane and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Denatured only)
  • Inhibits (40%) Binding of Proteoheparan to Smooth Muscle Cells
  • Inhibits Proliferation of Smooth Muscle Cells

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Moesin Ab-1
Monoclonal
Unconjugated
Mouse
1 mL
Cytoskeletal Protein
Bovine, Human, Rat
IgG2a κ
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
38/87
Ready-to-use for IHC (UVLP)
Moesin purified from cow uterus
RUO
Primary
Prediluted
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit