Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Moesin Ab-1, Mouse Monoclonal Antibody

Deliver accurate, reproducible results with the Thermo Scientific™ Moesin Ab-1, Mouse Monoclonal Antibody.

Specifications

Antigen Moesin Ab-1
Clone 38/87
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 1
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS727R1
View Documents
Thermo Scientific™
MS727R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Deliver accurate, reproducible results with the Thermo Scientific™ Moesin Ab-1, Mouse Monoclonal Antibody.

Moesin, a member of the talin-4.1 superfamily, is a linking protein of the submembraneous actin cytoskeleton. It is expressed in macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. It is involved in cell adhesion, migration, and organization of cell surface structures.

Host Species: Mouse

Clone: 38/87

Isotype: IgG1/κ

Species Reactivity: Human, Cow and Rat. Others not known.

Epitope: Not determined

Immunogen: Moesin purified from cow uterus

Molecular Weight: 78kDa moesin and 80kDa radixin

Positive Control: Raji cells; Placenta

Cellular Localization: Membrane and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Denatured only)
  • Inhibits (40%) Binding of Proteoheparan to Smooth Muscle Cells
  • Inhibits Proliferation of Smooth Muscle Cells

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Moesin Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Murine
Cytoskeletal Protein
Moesin purified from cow uterus
38/87
Monoclonal
RUO
Primary
IgG2a, κ
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit