Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Moexipril hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Angiotensin-converting enzyme (ACE) inhibitor that is hydrolyzed in the liver to the active metabolite moexiprilat (IC 50 values are 2.1 and 2700 nM for moexiprilat and moexipril respectively). Antihypertensive; decreases mean blood pressure in the spontaneous hypertensive rat (SHR). Also blocks degradation of bradykinin into inactive metabolites.
Specifications
Quantity | 10 mg |
Formula Weight | 535.03 |
Percent Purity | >99% |
Chemical Name or Material | Moexipril hydrochloride |
Safety and Handling
Recommended Storage | Desiccate at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction