Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MOGAT3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103518
Description
Antibody detects endogenous levels of total MOGAT3.
Catalyzes the formation of diacylglycerol from 2- monoacylglycerol and fatty acyl-CoA. Also able to catalyze the terminal step in triacylglycerol synthesis by using diacylglycerol and fatty acyl-CoA as substrates. Has a preference toward palmitoyl-CoA and oleoyl-CoA. May be involved in absorption of dietary fat in the small intestine by catalyzing the resynthesis of triacylglycerol in enterocytes.
Specifications
MOGAT3 | |
Polyclonal | |
Unconjugated | |
MOGAT3 | |
2-acylglycerol O-acyltransferase 3; acyl coenzyme A:monoacylglycerol acyltransferase 3; acyl-CoA:monoacylglycerol acyltransferase 3; DC7; DGAT2L2; DGAT2L7; Diacylglycerol acyltransferase 2-like protein 7; diacylglycerol O-acyltransferase candidate 7; hDC7; MGAT3; MOGAT3; Monoacylglycerol O-acyltransferase 3; UNQ9383/PRO34208 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
346606, 685560 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q86VF5 | |
MOGAT3 | |
A synthesized peptide derived from human MOGAT3(Accession Q86VF5), corresponding to amino acid residues H302-I341. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction