Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOGAT3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548315

Catalog No. PA548315

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Catalyzes the formation of diacylglycerol from 2- monoacylglycerol and fatty acyl-CoA. Also able to catalyze the terminal step in triacylglycerol synthesis by using diacylglycerol and fatty acyl-CoA as substrates. Has a preference toward palmitoyl-CoA and oleoyl-CoA. May be involved in absorption of dietary fat in the small intestine by catalyzing the resynthesis of triacylglycerol in enterocytes.
Specifications

Specifications

MOGAT3
Polyclonal
PBS with 0.09% sodium azide
Q86VF5
2-acylglycerol O-acyltransferase 3, Acyl-CoA:monoacylglycerol acyltransferase 3, MGAT3, Diacylglycerol O-acyltransferase candidate 7, hDC7, Diacylglycerol acyltransferase 2-like protein 7, Monoacylglycerol O-acyltransferase 3, MOGAT3, DC7, DGAT2L7
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
346606
Flow Cytometry, Western Blot
Unconjugated
MOGAT3
Liquid
MOGAT3
KLH conjugated synthetic peptide between 295-329 amino acids from the C-terminal region of human MOGT3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit