Moldex™ 1200N95 Series Dirt Dawgs Particulate Respirators
Disposable respirators with exclusive Moldex Dura-Mesh™ shell that protects the filter media so it stays cleaner-looking longer
Manufacturer: Moldex™ 1201N95
 View more versions of this product
Catalog No. 17384202
  Check Availability
Add to Cart
    Specifications
    Certifications ANSI ISEA 110-2003 7.11.1.
    Quantity 20
    Filter Type N95 Filter
    For Use With (Application) Hot, humid and dusty environments
    Includes Securely attached straps and naturally contoured facepiece
    Material Dura Mesh™
    Material (Liner) Softspun™
    Nosepiece Type Molded nose bridge
    Size Small
    Product Type Particulate Respirator
  • Description & Specifications
Related Products
Product Content Feedback