Learn More
Moldex™ 1200N95 Series Dirt Dawgs Particulate Respirators
Disposable respirators with exclusive Moldex Dura-Mesh™ shell that protects the filter media so it stays cleaner-looking longer
Manufacturer:  Moldex™ 1207N95
 View more versions of this product
Catalog No. 17384203
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
20
N95 Filter
Securely attached straps and naturally contoured facepiece
Softspun™
Low profile
Particulate Respirator
Hot, humid and dusty environments
Dura Mesh™
Molded nose bridge
ANSI ISEA 110-2003 7.11.1.