Learn More
Moldex™ 2360-P100 Particulate Respirator with Ventex™ Valve
All-day comfort with P100 protection
Manufacturer:  Moldex™ 2360AP100
 View more versions of this product
Catalog No. 19-384-347
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
1
White
Environments where an osha substance specific standard applies (lead, cadmium, arsenic, mda) as well as pharmaceutical manufacturing, conventional and nuclear power generation, welding, cutting and metal pouring
Dura Mesh™
ANSI ISEA 110-2003 7.11.1.
Particulate Respirator
P100 Filter
Ventex™ valve, soft foam nose cushion
Medium/Large