Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moldex™ EZ-ON™ N95 Particulate Respirators
SDP

First and only NIOSH N95 approved adjustable one-strap respirator.

Specifications

Aerosol Type N - Not Resistant to Oil
ANSI Standard ANSI/ISEA 110-2009, Section 7.11.1
Certifications (NIOSH) NIOSH 42 CFR 84, N95, approval number: TC-84A-3743
Color Code White
Design Type Cup/Molded
View More Specs

Products 2
Catalog Number Mfr. No. Size With Exhalation Valve Price Quantity & Availability  
Catalog Number Mfr. No. Size With Exhalation Valve Price Quantity & Availability  
19-149-720
SDP
View Documents
Moldex™
EZ22
Medium/Large No This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
19-149-723
SDP
View Documents
Moldex™
EZ23
Medium/Large Yes This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description

Description

This N95 respirator offers a quick fit for fast donning and doffing. Its one adjustable cloth strap with FLEXWING™ design feeds through a harness that secures the strap against the head and prevents slipping, snagging, and tangling.

Application

  • Hot, humid, and dusty environments where no oils are present, such as welding, soldering and brazing, grinding, sanding, sweeping, bagging, stone quarrying, metal machining, cement, underground mining, polishing, buffing, lime, poultry and textiles
  • Design
    • Dura-Mesh™ shell surrounds the mask's filter media, keeping it cleaner-looking longer; also helps respirator to hold its shape longer, extending service life
    • Design features no metal nose band to pinch closed; a soft foam nose cushion and contoured shape ensure a comfortable face seal
    • Available with or without a Ventex™ exhalation valve; valves help hot air to escape the mask faster, for cooler, more natural breathing

    Specifications

    Specifications

    N - Not Resistant to Oil
    NIOSH 42 CFR 84, N95, approval number: TC-84A-3743
    Cup/Molded
    N95
    General Purpose Particulate Respirator
    Welding, soldering and brazing, grinding, sanding, sweeping, bagging, stone quarrying, metal machining, cement, underground mining, polishing, buffing, lime, poultry and textiles
    10/Pk.
    No
    ANSI/ISEA 110-2009, Section 7.11.1
    White
    Yes
    Dura-Mesh™
    95%
    Molded Nose Bridge
    One Strap with Flexwing™
    SDS
    Product Certifications
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit