Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Moldex™ EZ-On™ Particulate Respirators
The first and only NIOSH N95 approved adjustable one-strap respirator
Manufacturer:  Moldex™ EZ 22
 View more versions of this product
Catalog No. 19-149-720
Add to cart
Specifications Description & Specifications
Specifications
10 Pack
95% or greater against non-oil based particulate aerosols
Yes
Welding, soldering and brazing, grinding, sanding, sweeping, bagging, stone quarrying, metal machining, cement, underground mining, polishing, buffing, lime, poultry and textiles
Aerosols free of oil
NIOSH TC-84A-3743, N95
E-Z On™ Particulate Respirator
White
N95 Filter
Dura Mesh™
Medium/Large