Moldex™ EZ-On™ Particulate Respirators
The first and only NIOSH N95 approved adjustable one-strap respirator
Manufacturer: Moldex™ EZ 22S
 View more versions of this product
Catalog No. 19-149-721
  Check Availability
Add to Cart
    Specifications
    Certifications NIOSH TC-84A-3743, N95
    Disposable Yes
    For Use With (Application) Welding, soldering and brazing, grinding, sanding, sweeping, bagging, stone quarrying, metal machining, cement, underground mining, polishing, buffing, lime, poultry and textiles
    Material Dura Mesh™
    Color Code White
    Filter Type N95 Filter
    Includes Ventex™ valve
    Product Type E-Z On™ Particulate Respirator
    Quantity 10 Pack
    Resistant To Aerosols free of oil
    Size Small
    Filter Efficiency 95% or greater against non-oil based particulate aerosols
  • Description & Specifications
Related Products
Product Content Feedback