Learn More
Moldex™ EZ-On™ Particulate Respirators
The first and only NIOSH N95 approved adjustable one-strap respirator
Manufacturer:  Moldex™ EZ 23
 View more versions of this product
Catalog No. 19-149-723
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
10 Pack
95% or greater against non-oil based particulate aerosols
Yes
Welding, soldering and brazing, grinding, sanding, sweeping, bagging, stone quarrying, metal machining, cement, underground mining, polishing, buffing, lime, poultry and textiles
Dura Mesh™
Medium/Large
E-Z On™ Particulate Respirator
White
N95 Filter
Ventex™ valve
Aerosols free of oil
NIOSH TC-84A-3746, N95