Learn More
Moldex™ HandyStrap™ Series Particulate Respirators
Buckle allows mask to hang around neck when not in use
Manufacturer:  Moldex™ 2601N95
 View more versions of this product
Catalog No. 19-039-694A
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
15 Pack
95% or greater against particulate aerosols free of oil
Yes
Grinding, sanding, sweeping, quarrying, machining and other dusty environments
Dura Mesh™
Non-oil based particles
ANSI ISEA 110-2003 7.11.1., NIOSH TC-84A-0013
Particulate Respirator
White
N95 Particulate Filter
Ventex™ valve and handystrap™
Softspun™
Small