Moldex™ HandyStrap™ Series Particulate Respirators

Buckle allows mask to hang around neck when not in use

Manufacturer: Moldex™ 2801N95

 View more versions of this product

Catalog No. 19-039-695A

  Check Availability
Add to Cart

    Specifications

    Certifications ANSI ISEA 110-2003 7.11.1., NIOSH TC-84A-3002
    Quantity 100 Case
    Disposable Yes
    Filter Type N95 Particulate Plus Nuisance Ozone and Organic Vapors Filter
    For Use With (Application) Welding applications, as well as other hot, humid, dusty environments
    Includes Ventex™ valve and handystrap™
    Material Dura Mesh™
    Material (Liner) Softspun™
    Size Small
    Product Type Particulate Respirator
    Filter Efficiency 95% or greater against particulate aerosols free of oil
    Color Gray

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit