Moldex™ HandyStrap™ Series Particulate Respirators
Buckle allows mask to hang around neck when not in use
Manufacturer: Moldex™ M2741R95
 View more versions of this product
Catalog No. 19-160-0167
  Check Availability
Add to Cart
    Specifications
    Certifications ANSI ISEA 110-2003 7.11.1., NIOSH
    Filter Type R95 Plus Ventex™ Valve Filter
    Material Dura Mesh™
    Size Small
    Quantity 10 Case
    Disposable Yes
    For Use With (Application) Welding, soldering and brazing, grinding, sanding, sweeping, bagging, stone quarrying, metal machining, cement, underground mining, polishing, buffing, lime, poultry and textiles
    Includes Ventex™ valve and handystrap™
    Nosepiece Type Soft foam nose cushion
    Resistant To Oil, non-oil based aerosol
    Product Type Particulate Respirator
    Filter Efficiency 95% or greater against particulate aerosols free of oil
    Color Black
  • Description & Specifications
Related Products
Product Content Feedback