Learn More
Moldex™ HandyStrap™ Series Particulate Respirators
Buckle allows mask to hang around neck when not in use
Manufacturer:  Moldex™ D2740R95
 View more versions of this product
Catalog No. 19-160-0168
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
10 Case
95% or greater against particulate aerosols free of oil
Yes
Welding, soldering and brazing, grinding, sanding, sweeping, bagging, stone quarrying, metal machining, cement, underground mining, polishing, buffing, lime, poultry and textiles
Dura Mesh™
Oil, non-oil based aerosol
ANSI ISEA 110-2003 7.11.1., NIOSH
Particulate Respirator
Tan
R95 Plus Ventex™ Valve Filter
Ventex™ valve and handystrap™
Soft foam nose cushion
Medium/Large