Moldex™ HandyStrap™ Series Particulate Respirators
Buckle allows mask to hang around neck when not in use
Manufacturer: Moldex™ 2941R95
 View more versions of this product
Catalog No. 19-999-368
  Check Availability
Add to Cart
    Specifications
    Certifications ANSI ISEA 110-2003 7.11.1., NIOSH TC-84A-3174
    Filter Type R95 Particulate Plus Nuisance Acid Gas Irritants Filter
    Includes Ventex™ valve and handystrap™
    Material Dura Mesh™
    Material (Liner) Softspun™
    Size Small
    Filter Efficiency 95% or greater against particulate aerosols free of oil
    Quantity 1 Pack
    Disposable Yes
    For Use With (Application) Use when oil aerosols are not present, including paper mills, aluminum reduction, coal-fired utilities and petrochemicals
    Resistant To Oil and non-oil based particles, nuisance levels of acid gas
    Product Type Particulate Respirator
    Color Gray
  • Description & Specifications
Related Products
Product Content Feedback