Learn More
Moldex™ HandyStrap™ Series Particulate Respirators Available on GSA/VA Contract for Federal Government customers only.
Buckle allows mask to hang around neck when not in use
Manufacturer:  Moldex™ 2731N100
 View more versions of this product
Catalog No. 19-047-826
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Disposable, maintenance-free design features convenient HandyStrap. Loosen the strap to allow the mask to hang freely around the neck when not in use
Application
  • See table matrix below
  • Design
    • HandyStrap feels cool and comfortable against skin
    • Strong Dura-Mesh™ shell resists collapsing in regular use and even under hot and humid conditions
    • Features comfortable contour-molded nose bridge with soft foam nose flange
    • Softspun™ lining enhances durability
    • 100% PVC free
    Styles
    • The 2600 N95 Series provides economical protection
    • All models (except 2600 N95) feature Ventex™ exhalation valve that reduces breathing resistance and vents hot, humid breath; workers breathe easier and stay cooler
    • The 2800 N95 and 2840 R95 Series include an added carbon layer to filter out nuisance levels of ozone and organic vapors (less than OSHA PEL)
    • The 2940 R95 Series protects against oil and oil-based particulates while filtering out nuisance levels of acid gas irritants (less than OSHA PEL)
    • Special Ops™ Models offer the same protection with a black Dura-Mesh shell
    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
    Certifications
    All are NIOSH certified. N95: 95% efficient, not for oil; N100: 99.97% efficient, not for oil; R95: 95% efficient, resistant to oil.
    Compliance
    Certified under NIOSH 42 CRF 84 to have a filter efficiencey level of at least 99.97% effective against particulate aerosols free of oil, up to 10X the OSHA Permissable Exposure Limit (PEL).
    Specifications
    Specifications
    Safety
    NIOSH 42 CRF 84
    95% or greater against particulate aerosols free of oil
    Pharmaceutical or battery manufacturing, welding and metal pouring
    Dura Mesh™
    Particulate Respirator
    Non-oil based particles
    White
    Yes
    N100 Particulate Plus Exhale valve Filter
    Ventex™ valve and handystrap™
    Softspun™
    5 Pack
    Small