Moldex™ N95 Series AirWave™ Particulate Respirators
Allows more airflow in and out for cool comfort and easier breathing
Manufacturer: Moldex™ 4201N95
 View more versions of this product
Catalog No. 19-999-372
  Check Availability
Add to Cart
    Specifications
    Certifications ANSI ISEA 110-2003 7.11.1.
    Quantity 10
    Filter Type N95 Filter
    For Use With (Application) Particulate aerosols free of oil
    Includes Pinch free nose bridge
    Material Dura Mesh™
    Material (Liner) Softspun™
    Nosepiece Type Soft foam nose cushion
    Size Small
    Product Type Disposable Respirator
    Design Type Flexfit
  • Description & Specifications
Related Products
Product Content Feedback