Moldex™ PlugStation™ Ear Plug Dispenser

Wall-mountable dispenser puts ear plugs in easy reach

Manufacturer: Moldex™ 6645

 View more versions of this product

Catalog No. 19-003-410B

  Check Availability
Add to Cart

    Specifications

    Cord Type Uncorded
    Includes SparkPlugs™ ear plugs; Wall mount hardware
    Material Foam
    Noise Reduction Range (NRR) 33dB
    Quantity 4 Case
    Weight (Metric) 0.02kg
    Size Large
    Class 5
    Color Assorted
    Disposable Yes
    Holds 500 Pairs
    Shape Tapered
    Weight (English) 0.05 lb.
    Product Type Earplug Dispenser

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit