Moldex™ Rockets™ Reusable Ear Plugs

NRR: 27dB

Manufacturer: Moldex™ 6420

 View more versions of this product

Catalog No. 19-063-065

  Check Availability
Add to Cart

    Specifications

    Product Type Rockets™ Earplug
    Shape Flange
    Style With optional Cloth cord
    Noise Reduction Range (NRR) 27dB
    Certifications/Compliance ANSI S3.19-1974
    Size One size fits to all
    Color Purple
    Disposable No
    For Use With (Application) Soft flanges fit most ear canals and provide a superior seal

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit