Moldex™ SparkPlugs™ Foam Earpugs

Tapered shape earplugs that are comfortable and offer a high NRR 33 rating.

Manufacturer: Moldex™ 6604

 View more versions of this product

Catalog No. 18-999-2271

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Most noisy environments
    Product Type Sparkplugs™ Earplug
    Material Polyurethane Foam
    Shape Tapered
    Style Uncorded
    Noise Reduction Range (NRR) 33dB
    Certifications/Compliance ANSI S3.19-1974
    Size One size fits to all
    Color Multicolor
    Disposable Yes

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit